2,6-Dichloropyridine-4-methanol 95+%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
2,6-Dichloro-4-pyridinemethanol, 2,6-Dichloro-4-(hydroxymethyl)pyridine
OR0150-1G
OR0150-5G
76615-148EA
209.09
USD
76615-148
76615-150
76615-152
2,6-Dichloropyridine-4-methanol 95+%
2,6-Dichloropyridine-4-methanol
Formula:
C₆H₅Cl₂NO Melting Pt: 127…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052638 CAS Number: 101990-69-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...