Copper(I) iodide 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Cuprous iodide

IN1559-100G IN1559-1KG IN1559-500G
76615-078EA 23.44 USD
76615-078 76615-080 76615-082
Copper(I) iodide 98%
Copper (I) iodide
Formula: CuI
Boiling Pt: 1290 °C
Melting Pt: 605 °C
Density: 5.62 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR