Copper(II) bromide, Reagent Grade

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Cupric bromide, Cuprous bromide

IN1540-100G
76615-076EA 30.56 USD
76615-076
Copper(II) bromide, Reagent Grade
Copper (II) bromide
Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900 °C
Melting Pt: 498 °C
Density: 4.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR