PA43 BIO Trinocular Ergonomic Pathology Lab Microscpe Packages, Motic
Supplier: Motic
PA43PATHE-S6
76615-056EA 9622 USD
76615-056 76615-058 76615-060
PA43 BIO Trinocular Ergonomic Pathology Lab Microscpe Packages, Motic
Microscopes
The new PA43 BIOMED is a new series from Motic scientific and represents a large step forward in combining modularity with ergonomics to improve application imaging workflows. A progression into routine work, this new microscope’s robust design and solid footprint does not compromise on optical quality and features.
  • PA43BIO trinocular ergonomic upright pathology lab microscope
  • 0.5× C-mount camera adapter for ¹/₃" and ¹/₂" chip sensors
  • Motic analyzes basic 64-bit; USB + dongle (no charge)
  • Moticam S6/1080X/BMH4000
Its modularity offers flexibility when choosing a configuration that suits multiple end-user applications from cost-effective solutions to high-quality optical systems for lab diagnostics. The microscope is not only modular but also flexible in design and can be upgraded at any time.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.