AE2000MET Inverted Materials Trinocular Microscopes with 100 W Illumination, Motic
Supplier: Motic
MB-AE2000MT MB-AE2000MT-BF
76614-978EA 10032.26 USD
76614-978 76614-980
AE2000MET Inverted Materials Trinocular Microscopes with 100 W Illumination, Motic
Microscopes
Motic’s AE2000MET is an inverted metallurgical microscope designed to inspect polished cross-sections as well as larger, bulky samples within industries such as automotive, machinery, and metal fabrication.
  • Improved ergonomy for efficient work
  • Smart phase contrast for daily routine work
  • A powerful and flexible illumination concept
  • Phase contrast for unstained samples
  • Reproducible documentation results with high reliability
Motic’s CCIS® advanced Infinity optics provide brightfield/darkfield objectives with long working distances, while quality glass and anti-reflex coating ensures superb image quality. In addition to the standard package, the AE2000MET offers a polarizer and analyzer for cross polarization. By combining the AE2000MET with a Moticam option, advanced measurement, image processing and documentation-reporting is available through the Motic analyzes software.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty