PA53 MET Trinocular Upright Materials Microscope - 100 W Reflected Illumination, Motic
Supplier: Motic
1100104600851
76614-942EA 10016.84 USD
76614-942
PA53 MET Trinocular Upright Materials Microscope - 100 W Reflected Illumination, Motic
Microscopes
Motic new PA53 MET microscopes introduce a superior optical performance, implemented in an ergonomic hardware setup, and combined with automated functions for precise quantifications.
  • Expertly designed, carefully crafted, high-performance Motic objective lenses provide excellent clarity in any observation environment
  • Advanced ergonomic design, compact size, carefully sourced high-quality materials, and versatile digital capabilities ensure minimal operator fatigue during extended periods of operation
  • Clear and vivid images are provided through objective
  • lenses and illuminators of the finest quality which have
  • been designed with aspheric lenses
The Motic PA53 MET Series: Ultimate solutions for industrial quality control and analyzes of opaque and transparent materials.
Ordering information: Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.