Motic BA310 Metallurgical Microscope
Supplier: VWR International
1100100402144 1100100402045 1100100402601 1100100402524 1100100403591
10767-558EA 4509.62 USD
10767-558 10767-560 76163-494 76163-628 76614-920
Motic BA310 Metallurgical Microscope
Microscopes
The Motic BA310Met Advanced Metallurgical microscope series is designed to meet the performance demands of engineering professionals who study opaque samples like minerals, metals, and building materials.
The specialized M Plan achromatic metallurgical objectives (5x, 10x, 20x, 50x) are set in a reversed quintuple nosepiece, providing faithful reproduction of samples up to 30mm in thickness
The BA310Met features a Siedentopf style head inclined at 30° with full 360° rotation for pain-free viewing over extended periods of time. The M Plan Achromatic lenses provide improved working distance (8.2mm at 500x), allowing for sample manipulations while viewing. Additional add-ons such as a polarizer or the Moticam series of microscopy cameras further enhance the BA310Met for metallurgical observation and documentation. The BA310Met provides a robust, reliable tool for education, engineering and material science, industrial quality control, and many other material applications.
The standard halogen epi-illuminator includes adjustable field and aperture diaphragms to minimize reflections and optimize performance. Conversion of the standard halogen illumination to a 3W LED is easily achieved with a simple plug-in module.
Ordering information: Microscope is supplied with all accessories listed above and micrometer reticles. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.