2-Bromo-1-ethylpyridinium tetrafluoroborate 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
BEP, 2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate
PC8735-100G
PC8735-25G
PC8735-5G
76614-664EA
321.9
USD
76614-664
76614-666
76614-668
2-Bromo-1-ethylpyridinium tetrafluoroborate 98%
2-Bromo-1-ethylpyridinium tetrafluoroborate
Formula:
C₇H₉BBrF₄N Melting Pt: 100…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060046 CAS Number: 878-23-9 EINECS: 212-900-2 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...