2-Bromo-5-fluorostyrene

Supplier: APOLLO SCIENTIFIC
Danger

PC7261-1G PC7261-250MG
76614-510EA 132.12 USD
76614-510 76614-512
2-Bromo-5-fluorostyrene
2-Bromo-5-fluorostyrene
Formula: C₈H₆BrF
Storage Temperature: Refrigerator
MDL Number: MFCD09749661
CAS Number: 828267-47-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR