(Bromodifluoromethyl)trimethylsilane 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC53035-1G
PC53035-25G
PC53035-5G
76614-234EA
34.64
USD
76614-234
76614-236
76614-238
(Bromodifluoromethyl)trimethylsilane 97%
(Bromodifluoromethyl)trimethylsilane
Formula:
C₄H₉BrF₂Si Boiling Pt: 112 °C Density: 1.41 g/cm³ (25 °C) Flash Pt: 45 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18641931 CAS Number: 115262-01-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...