(Bromodifluoromethyl)trimethylsilane 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC53035-1G PC53035-25G PC53035-5G
76614-234EA 34.64 USD
76614-234 76614-236 76614-238
(Bromodifluoromethyl)trimethylsilane 97%
(Bromodifluoromethyl)trimethylsilane
Formula: C₄H₉BrF₂Si
Boiling Pt: 112 °C
Density: 1.41 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18641931
CAS Number: 115262-01-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR