4-Fluoro-3-nitroaniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3880-100G PC3880-25G PC3880-500G
76613-812EA 23.44 USD
76613-812 76613-814 76613-816
4-Fluoro-3-nitroaniline 98%
4-Fluoro-3-nitroaniline
Formula: C₆H₅FN₂O₂
Melting Pt: 94…98 °C
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007833
CAS Number: 364-76-1
EINECS: 206-665-5
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR