4-Amino-1,1,1-trifluorobut-3-en-2-one

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Amino-2-oxo-1,1,1-trifluorobut-3-ene, 4-Amino-1,1,1-trifluoro-3-buten-2-one

PC31145-1G
76613-710EA 82.52 USD
76613-710
4-Amino-1,1,1-trifluorobut-3-en-2-one
4-Amino-1,1,1-trifluorobut-3-en-2-one
Formula: C₄H₄F₃NO
MW: 139.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01569588
CAS Number: 184848-89-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR