4-Bromo-2-chloro-6-(trifluoromethyl)benzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC

PC303390-1G PC303390-250MG
76613-642EA 847.54 USD
76613-642 76613-644
4-Bromo-2-chloro-6-(trifluoromethyl)benzenesulfonyl chloride 97%
4-Bromo-2-chloro-6-(trifluoromethyl)benzenesulfonyl chloride
Formula: C₇H₂BrCl₂F₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD31916398
CAS Number: 2384019-85-4
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR