1-Bromo-3,4,5-trifluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-Bromo-1,2,3-trifluorobenzene

PC1575-100G PC1575-25G
76612-854EA 41.76 USD
76612-854 76612-856
1-Bromo-3,4,5-trifluorobenzene 99%
1-Bromo-3,4,5-trifluorobenzene
Formula: C₆H₂BrF₃
MW: 210.98
Boiling Pt: 140
Melting Pt: -20
Density: 1.767 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00070738
CAS Number: 138526-69-9
EINECS: 418-480-9
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR