2,3,5-Trifluoropyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC10551-100G PC10551-25G PC10551-5G
76612-732EA 388.12 USD
76612-732 76612-734 76612-736
2,3,5-Trifluoropyridine 98%
2,3,5-Trifluoropyridine
Formula: C₅H₂F₃N
Boiling Pt: 102 °C
Density: 1.4 g/cm³ (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD03001162
CAS Number: 76469-41-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR