4-Amino-2,2,6,6-tetramethylpiperidine 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR6937-100G
OR6937-500G
76612-360EA
58.06
USD
76612-360
76612-362
76612-364
4-Amino-2,2,6,6-tetramethylpiperidine 98%
4-Amino-2,2,6,6-tetramethylpiperidine
Formula:
C₉H₂₀N₂ Boiling Pt: 188…189 °C Melting Pt: 16…18 °C Density: 0.912 g/cm³ (25 °C) Flash Pt: 75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005984 CAS Number: 36768-62-4 EINECS: 253-197-2 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...