1-Allylpiperazine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR6900-1G OR6900-5G
76612-336EA 39.26 USD
76612-336 76612-338
1-Allylpiperazine 97%
1-Allylpiperazine
Formula: C₇H₁₄N₂
Boiling Pt: 180…182 °C
Density: 0.902 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00167970
CAS Number: 13961-36-9
UN: 2734
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR