4,4-Dimethoxybutanone

Supplier: APOLLO SCIENTIFIC
Warning

OR59432-5G
76612-158EA 20.87 USD
76612-158
4,4-Dimethoxybutanone
4,4-Dimethoxybutanone
Formula: C₆H₁₂O₃
Boiling Pt: 170…172 °C
Density: 0.996 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00008789
CAS Number: 5436-21-5
EINECS: 226-605-1
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR