2-Bromo-N,N-dimethylacetamide 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR55573-25G OR55573-5G
76612-130EA 135.48 USD
76612-130 76612-132
2-Bromo-N,N-dimethylacetamide 98%
2-Bromo-N,N-dimethylacetamide
Formula: C₄H₈BrNO
Boiling Pt: 63…65 °C/1mmHg
Density: 1.5412 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD12026683
CAS Number: 5468-77-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR