Allylboronic acid pinacol ester 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

OR3740-100G OR3740-25G OR3740-5G
76611-762EA 173.18 USD
76611-762 76611-764 76611-766
Allylboronic acid pinacol ester 97%
Allylboronic acid pinacol ester
Formula: C₉H₁₇BO₂
MW: 168.05 g/mol
Boiling Pt: 50…53 °C/5mmHg
Density: 0.896 g/cm³ (25 °C)
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00013347
CAS Number: 72824-04-5
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR