p-Toluenesulfonic acid monohydrate 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate, 4-Methylbenzenesulfonic acid hydrate, Toluene-4-sulfonic acid monohydrate

OR322135-100G
76611-694EA 21.36 USD
76611-694 76611-696
p-Toluenesulfonic acid monohydrate 99%
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
Boiling Pt: 140 °C/20mmHg
Melting Pt: 103…106 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
UN: 2585
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR