Benzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Aminotoluene

OR16591-1KG OR16591-250G OR16591-5KG
76611-210EA 82.52 USD
76611-210 76611-212 76611-214
Benzylamine 98%
Benzylamine
Formula: (C₆H₅)CH₂NH₂
Boiling Pt: 183…185 °C
Melting Pt: 10 °C
Density: 0.981 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00008106
CAS Number: 100-46-9
EINECS: 202-854-1
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR