Benzylamine 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
α-Aminotoluene
OR16591-1KG
OR16591-250G
OR16591-5KG
76611-210EA
82.52
USD
76611-210
76611-212
76611-214
Benzylamine 98%
Benzylamine
Formula:
(C₆H₅)CH₂NH₂ Boiling Pt: 183…185 °C Melting Pt: 10 °C Density: 0.981 g/cm³ (25 °C) Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008106 CAS Number: 100-46-9 EINECS: 202-854-1 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...