N-Boc-ethylenediamine 97+%

Supplier: APOLLO SCIENTIFIC
Danger

OR1533-100G OR1533-25G
76611-152EA 55.02 USD
76611-152 76611-154 76611-156
N-Boc-ethylenediamine 97+%
N-Boc-ethylenediamine
Formula: C₇H₁₆N₂O₂
Boiling Pt: 72…80 °C/0.1mmHg
Density: 1.012 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00191871
CAS Number: 57260-73-8
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR