Benzoxazole

Supplier: APOLLO SCIENTIFIC
Warning

OR13697-25G
76611-112EA 20.28 USD
76611-112
Benzoxazole
Benzoxazole
Formula: C₇H₅NO
Boiling Pt: 181…183 °C
Melting Pt: 27…31 °C
Density: 1.175 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005765
CAS Number: 273-53-0
EINECS: 205-988-9
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR