Calcium nitrate tetrahydrate

Supplier: APOLLO SCIENTIFIC
Danger

OR13216-2.5KG OR13216-100G OR13216-1KG
76611-088EA 93.72 USD
76611-088 76611-084 76611-086
Calcium nitrate tetrahydrate
Calcium nitrate tetrahydrate
Formula: Ca(NO₃)₂·4H₂O
Boiling Pt: 132 °C
Melting Pt: 40…45 °C
Density: 1.82 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR