(±)-3-Amino-1,2-propanediol 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR10134-100G OR10134-25G OR10134-500G
76610-920EA 48.9 USD
76610-920 76610-922 76610-924
(±)-3-Amino-1,2-propanediol 98%
(±)-3-Amino-1,2-propanediol
Formula: C₃H₉NO₂
Boiling Pt: 264…265 °C
Density: 1.175 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008140
CAS Number: 616-30-8
EINECS: 210-475-8
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR