2-Aminopyridine 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR0015-1KG
76610-808EA 101.88 USD
76610-808 76610-810
2-Aminopyridine 99%
2-Aminopyridine
Formula: C₅H₆N₂
Boiling Pt: 204…211 °C
Melting Pt: 54…60 °C
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006312
CAS Number: 504-29-0
EINECS: 207-988-4
UN: 2671
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR