Tin(IV) chloride hydrate 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Stannic chloride, Tin tetrachloride hydrate

IN3650-25G
76610-802EA 82.4 USD
76610-802 76610-804 76610-806
Tin(IV) chloride hydrate 98%
Tin(IV) chloride hydrate
Formula: SnCl₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149864
CAS Number: 7646-78-8
UN: 2440
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR