2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-5-(trifluoromethyl)isonicotinic acid

PC8968-10G PC8968-1G PC8968-5G
76609-750EA 90.64 USD
76609-750 76609-752 76609-754
2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid 98%
2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
Formula: C₇H₃ClF₃NO₂
Melting Pt: 185…186 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01862019
CAS Number: 505084-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR