7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC8617-1G
76609-474EA 54.72 USD
76609-474
7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
Formula: C₁₀H₅F₃O₂S
Melting Pt: 191…195 °C
Storage Temperature: Ambient
MDL Number: MFCD09027110
CAS Number: 550998-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR