3-Cyano-2-fluorophenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC8333-1G
76609-358EA 33.62 USD
76609-358
3-Cyano-2-fluorophenylboronic acid
3-Cyano-2-fluorophenylboronic acid
Formula: C₇H₅BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD07374984
CAS Number: 957121-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR