1-(3,5-Bis(trifluoromethyl)phenyl)biguanide hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

PC7824-100G PC7824-25G PC7824-5G
76608-936EA 1256.54 USD
76608-936 76608-938 76608-940
1-(3,5-Bis(trifluoromethyl)phenyl)biguanide hydrochloride
1-(3,5-Bis(trifluoromethyl)phenyl)biguanide hydrochloride
Formula: C₁₀H₁₀ClF₆N₅
Melting Pt: 246 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03094450
CAS Number: 36068-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR