Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate

Supplier: APOLLO SCIENTIFIC
Warning

PC7795-100G PC7795-25G PC7795-5G
76608-852EA 77.1 USD
76608-852 76608-854 76608-856
Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate
Ethyl (2,3,4,5-tetrafluorobenzoyl)acetate
Formula: C₁₁H₈F₄O₃
Boiling Pt: 100 °C/0.075mmHg
Melting Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00523020
CAS Number: 94695-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR