1-Nitro-4-(trifluoromethoxy)benzene 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Trifluoromethoxy)nitrobenzene

PC7439K-100G PC7439K-25G PC7439K-5G
76608-552EA 46.86 USD
76608-552 76608-554 76608-556
1-Nitro-4-(trifluoromethoxy)benzene 97%
1-Nitro-4-(trifluoromethoxy)benzene
Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Boiling Pt: 87…89 °C/15mmHg
Melting Pt: 15 °C
Density: 1.447 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040302
CAS Number: 713-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR