6-(Trifluoromethyl)isatoic anhydride 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC500009-1G PC500009-250MG
76606-156EA 304.58 USD
76606-156 76607-304
6-(Trifluoromethyl)isatoic anhydride 97%
6-(Trifluoromethyl)isatoic anhydride
Formula: C₉H₄F₃NO₃
Melting Pt: 239 °C
Storage Temperature: Ambient
MDL Number: MFCD24850066
CAS Number: 387-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR