2-Fluoro-4-methyl-5-nitroanisole 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC500308-1G PC500308-250MG
76603-934EA 462.48 USD
76603-934 76607-158
2-Fluoro-4-methyl-5-nitroanisole 95%
2-Fluoro-4-methyl-5-nitroanisole
Formula: C₈H₈FNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD24368898
CAS Number: 3108-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR