3-Bromo-4-fluoro-2-methoxyaniline

Supplier: APOLLO SCIENTIFIC
Warning

PC49008-1G PC49008-250MG
76605-506EA 261.28 USD
76605-506 76606-372
3-Bromo-4-fluoro-2-methoxyaniline
3-Bromo-4-fluoro-2-methoxyaniline
Formula: C₇H₇BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD18071064
CAS Number: 1257535-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR