5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC4924-100MG PC4924-1G
76604-032EA 69.26 USD
76604-032 76606-300
5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
Formula: C₁₀H₇F₃N₂O₂S
Melting Pt: 212…215 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00206674
CAS Number: 223499-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR