4-(Bromomethyl)-2-fluorobenzeneboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC500756-1G PC500756-5G
76607-656EA 247.54 USD
76607-656 76604-590
4-(Bromomethyl)-2-fluorobenzeneboronic acid
4-(Bromomethyl)-2-fluorobenzeneboronic acid
Formula: C₇H₇BBrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD22573054
CAS Number: 1331945-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR