3-Amino-8-fluoroquinoline dihydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

PC49034-1G PC49034-250MG
76604-478EA 349.48 USD
76604-478 76605-272
3-Amino-8-fluoroquinoline dihydrochloride
3-Amino-8-fluoroquinoline dihydrochloride
Formula: C₉H₇FN₂·2ClH
Storage Temperature: Ambient
MDL Number: MFCD18375334
CAS Number: 1266226-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR