4-Bromo-3-(trifluoromethoxy)benzoic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC49243-1G PC49243-250MG
76604-220EA 303.56 USD
76604-220 76604-292
4-Bromo-3-(trifluoromethoxy)benzoic acid 97%
4-Bromo-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₄BrF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD22378818
CAS Number: 1008769-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR