2-Fluoro-4-hydroxy-5-nitrobenzaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC500450-1G PC500450-250MG PC500450-5G
76604-118EA 166.04 USD
76604-118 76606-916 76607-532
2-Fluoro-4-hydroxy-5-nitrobenzaldehyde 95%
2-Fluoro-4-hydroxy-5-nitrobenzaldehyde
Formula: C₇H₄FNO₄
Storage Temperature: Ambient
MDL Number: MFCD27995610
CAS Number: 394-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR