1H,1H,2H,2H-Tridecafluoro-1-octanol 97+%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctan-1-ol, 1H,1H,2H,2H-Perfluoro-1-octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanol, 1H,1H,2H,2H-Perfluorooctan-1-ol, 1H,1H,2H,2H-Perfluorooctanol
PC6147-25G
PC6147-100G
PC6147-250G
76603-508EA
20.99
USD
76603-508
76603-504
76603-506
1H,1H,2H,2H-Tridecafluoro-1-octanol 97+%
1H,1H,2H,2H-Tridecafluoro-1-octanol
Formula:
C₈H₅F₁₃O MW: 364.11 g/mol Boiling Pt: 91…93 °C/36mmHg Density: 1.678 g/cm³ (25 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042143 CAS Number: 647-42-7 EINECS: 211-477-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...