Pentafluorophenol 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC5630-100G PC5630-500G
76602-458EA 33.62 USD
76602-458 76602-460
Pentafluorophenol 99%
Pentafluorophenol

Useful reagent for peptide coupling via the reactive PFP esters, often prepared by carbodiimide coupling.

Formula: C₆HF₅O
Boiling Pt: 143 °C
Melting Pt: 34…36 °C
Density: 1.757 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002156
CAS Number: 771-61-9
EINECS: 212-235-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR