1-(1,1-Difluoroethyl)-2-fluorobenzene 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC56077-1G
PC56077-250MG
76602-212EA
125.3
USD
76602-212
76602-214
1-(1,1-Difluoroethyl)-2-fluorobenzene 98%
1-(1,1-Difluoroethyl)-2-fluorobenzene
Formula:
C₈H₇F₃ Storage Temperature: Ambient |
MDL Number:
MFCD11845814 CAS Number: 1138445-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...