1-(1,1-Difluoroethyl)-2-fluorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC56077-1G PC56077-250MG
76602-212EA 125.3 USD
76602-212 76602-214
1-(1,1-Difluoroethyl)-2-fluorobenzene 98%
1-(1,1-Difluoroethyl)-2-fluorobenzene
Formula: C₈H₇F₃
Storage Temperature: Ambient
MDL Number: MFCD11845814
CAS Number: 1138445-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR