4'-Bromo-2',5'-difluoroacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

PC56074-1G PC56074-250MG PC56074-5G
76602-206EA 23.34 USD
76602-206 76602-208 76602-210
4'-Bromo-2',5'-difluoroacetophenone
4'-Bromo-2',5'-difluoroacetophenone
Formula: C₈H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD15526896
CAS Number: 123942-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR