2,4,6-Trifluorophenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC5536-1G PC5536-25G PC5536-5G
76602-070EA 20.32 USD
76602-070 76602-072 76602-074
2,4,6-Trifluorophenylboronic acid
2,4,6-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
Melting Pt: 228…235 °C
Storage Temperature: Ambient
MDL Number: MFCD01863169
CAS Number: 182482-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR